Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM69B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565871
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565871 Supplier Invitrogen™ Supplier No. PA565871
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EELGTQLRTCT TLSGLASQVE AHHSLVLSHL KTLLWK Highest antigen sequence identity to the following orthologs - mouse 92%, rat 95%.

This gene encodes a member of the FAM69 family of cysteine-rich type II transmembrane proteins. These proteins localize to the endoplasmic reticulum but their specific functions are unknown. [provided by RefSeq, Nov 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM69B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM69B
Gene Accession No. Q5VUD6
Gene Alias AB030186; AW209000; B230317C12Rik; C9orf136; DIPK1B; Divergent protein kinase domain 1B; Fam69b; family with sequence similarity 69 member B; family with sequence similarity 69, member B; pancreatitis-induced protein 49; PIP49; pp6977; Protein FAM69B
Gene Symbols FAM69B
Host Species Rabbit
Immunogen Recombinant Human FAM69B. Recombinant protein control fragment (Product #RP-106542).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138311
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.