Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM71A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569922
Description
This target displays homology in the following species: Human: 100%.
This gene encodes a protein that interacts with the Rab2B small GTPase and may be important for integrity of the Golgi body. A knockdown of this gene induces fragmentation of the Golgi, similar to the effect seen with a knockdown of the Rab2B small GTPase. The encoded protein has an N-terminal Rab-binding domain specific for Rab2B.
Specifications
FAM71A | |
Polyclonal | |
Unconjugated | |
FAM71A | |
FAM71A; family with sequence similarity 71 member A; family with sequence similarity 71, member A; protein FAM71A; testis secretory sperm-binding protein Li 209a | |
Rabbit | |
Affinity Chromatography | |
RUO | |
149647 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IYT1 | |
FAM71A | |
synthetic peptide directed towards the C terminal region of human FAM71A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction