Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM71A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555864
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555864 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555864 Supplier Invitrogen™ Supplier No. PA555864
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SMSLSREGSV SLAIAGVVLT SRTAAEADMD AAAGPPVSTR QSKSSLSGQH GRERTQASAE GCKEGRERRE KDRALGRSSH RRRTGESRHK Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 38%.

This gene encodes a protein that interacts with the Rab2B small GTPase and may be important for integrity of the Golgi body. A knockdown of this gene induces fragmentation of the Golgi, similar to the effect seen with a knockdown of the Rab2B small GTPase. The encoded protein has an N-terminal Rab-binding domain specific for Rab2B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM71A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM71A
Gene Accession No. Q8IYT1
Gene Alias FAM71A; family with sequence similarity 71 member A; family with sequence similarity 71, member A; protein FAM71A; testis secretory sperm-binding protein Li 209a
Gene Symbols FAM71A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM71A. Recombinant protein control fragment (Product #RP-94408).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149647
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.