Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM71B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | FAM71B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
FAM71B Polyclonal specifically detects FAM71B in Human samples. It is validated for Western Blot.Specifications
FAM71B | |
Polyclonal | |
Rabbit | |
Q8TC56 | |
153745 | |
Synthetic peptides corresponding to FAM71B(family with sequence similarity 71, member B) The peptide sequence was selected from the middle region of FAM71B. The immunogen falls within the following peptide sequence: RREKKDRHPSRKSSHHRKAGESHRRRAGDKNQKASSHRSASGHKNTRDDK. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
family with sequence similarity 71, member B, MGC26988 | |
FAM71B | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title