Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM71C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113227
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113227 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113227 Supplier Invitrogen™ Supplier No. PA5113227
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM71C Polyclonal Antibody for Western Blot, ELISA

FAM71C is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM71C
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FAM71C
Gene Accession No. Q8NEG0
Gene Alias FAM71C; family with sequence similarity 71 member C; family with sequence similarity 71, member C; protein FAM71C
Gene Symbols FAM71C
Host Species Rabbit
Immunogen Recombinant Human Protein FAM71C protein (1-241AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196472
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.