Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM71D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598619
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598619 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598619 Supplier Invitrogen™ Supplier No. PA598619
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM71D Polyclonal Antibody for Western Blot, ELISA

FAM71D is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM71D
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FAM71D
Gene Accession No. Q8N9W8
Gene Alias 4921509E07Rik; 4930516C23Rik; C14orf54; Fam71d; family with sequence similarity 71 member D; family with sequence similarity 71, member D; GARI-L2; Golgi-associated Rab2B interactor-like 2; protein FAM71D
Gene Symbols FAM71D
Host Species Rabbit
Immunogen Recombinant Human Protein FAM71D protein (220-398AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161142
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.