Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM71F1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144170
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144170 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144170 Supplier Invitrogen™ Supplier No. PA5144170
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM71F1 Polyclonal Antibody for IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM71F1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene FAM71F1
Gene Accession No. Q96KD3
Gene Alias FAM137A; FAM71F1; family with sequence similarity 137, member A; family with sequence similarity 71 member F1; family with sequence similarity 71, member F1; NYD-SP18; Protein FAM137A; Protein FAM71F1; testes development-related NYD-SP18; testicular tissue protein Li 67; testis development protein NYD-SP18
Gene Symbols FAM71F1
Host Species Rabbit
Immunogen Recombinant Human Protein FAM71F1 protein (121-343AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84691
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.