Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM71F1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310224100UL
Description
FAM71F1 Polyclonal specifically detects FAM71F1 in Mouse samples. It is validated for Western Blot.Specifications
FAM71F1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
FAM137A, family with sequence similarity 71, member F1, member A, Protein FAM137A, Testis development protein NYD-SP18 | |
The immunogen is a synthetic peptide directed towards the middle region of mouse FAM71F1 (NP_001276592.1). Peptide sequence MVHSPEFNLFPNSVVFESNFVQVRRSRNWKEIYKASNTMALGVTSSVPCL | |
100 μg | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
84691 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction