Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ FAM71F2 Polyclonal Antibody
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555026
Description
Immunogen sequence: ADRNNDTAIE IDNCSSYKIP SPVASPINLN IPMRAALSHS LWEQEDWNEH LLQVHIASYL GEHFLGA Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 57%.
            Specifications
| FAM71F2 | |
| Polyclonal | |
| Unconjugated | |
| Fam71f2 | |
| AI507611; FAM137B; FAM71F2; family with sequence similarity 137, member B; family with sequence similarity 71 member F2; family with sequence similarity 71, member F2; GARI; Golgi-associated Rab2B interactor; Protein FAM137B; Protein FAM71F2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 346653 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | 
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NXP2 | |
| Fam71f2 | |
| Recombinant protein corresponding to Human FAM71F2. Recombinant protein control fragment (Product #RP-102545). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction