Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM72 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501733316 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3316 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3316 Supplier Proteintech Group Inc Supplier No. 212031AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects all the members of FAM72.

FAM72A, also named as LMIP and UGENE, is a mitochondrial protein which regulates cellular reactive oxygen species metabolism. It is up-regulated in maligt colon cancers, compared to normal colon and colon adenomas. FAM72 has some members A/B/C/D with higher homology.

Specifications

Antigen FAM72
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FAM72A
Gene Accession No. A1KXW8, Q5TYM5, Q8BFZ8
Gene Alias FAM72, FAM72A, FAM72B, FAM72C, FAM72D, Protein FAM72A
Gene Symbols FAM72A
Host Species Rabbit
Immunogen FAM72 Fusion Protein Ag15566
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108900, 681249, 729533
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.