Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM72A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583650 Supplier Invitrogen™ Supplier No. PA583650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CVSILCCKFC KQVLSSRGMK AVLLADTEID LFSTDIPPTN AVDFTGRCYF T Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 94%.

FAM72A, also named as LMIP and UGENE, is a mitochondrial protein which regulates cellular reactive oxygen species metabolism. It is up-regulated in malignant colon cancers, compared to normal colon and colon adenomas. FAM72 has some members A/B/C/D with higher homology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM72A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM72A
Gene Accession No. Q5TYM5
Gene Alias 2700049P18Rik; dpr4-related protein; Fam72a; family with sequence similarity 72 member A; family with sequence similarity 72, member A; latent membrane protein 1-induced protein; LMP1-induced protein; LMPIP; MGC57827; P17; Protein FAM72A; RP11-312O7.1; Ugene
Gene Symbols FAM72A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM72A. Recombinant protein control fragment (Product #RP-108173).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729533
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.