Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM73B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571096 Supplier Invitrogen™ Supplier No. PA571096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rat: 100%.

MIGA2 gene ontology annotations related to this gene include mitochondrial fusion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM73B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MIGA2
Gene Accession No. Q7L4E1
Gene Alias 5730472N09Rik; AA408683; AI790341; C9orf54; FAM73B; family with sequence similarity 73 member B; family with sequence similarity 73, member B; hypothetical protein LOC84895; mFLJ00199 protein; MIGA2; Mitoguardin 2; mitoguardin-2; ortholog of family with sequence similarity 73, member B FAM73B; protein FAM73B; PSEC0112; R74766; RGD1310887
Gene Symbols MIGA2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human FAM73B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84895
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.