Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM76A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989236
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9236 Supplier Bioss Supplier No. BS9658R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM76A is a protein coding gene.

Specifications

Antigen FAM76A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Fam76a
Gene Accession No. Q8TAV0
Gene Alias FAM76A; family with sequence similarity 76 member A; family with sequence similarity 76, member A; Protein FAM76A; RGD1305162
Gene Symbols Fam76a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM76A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199870
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.