Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM76B Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178757F
Description
FAM76B Polyclonal antibody specifically detects FAM76B in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
FAM76B | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
family with sequence similarity 76, member B, hypothetical protein LOC143684 | |
The immunogen for this product maps to a region between residue 289 and 339 of human Family with Sequence Similarity 76, Member B using the numbering given in entry NP_653265.3 (GeneID 143684). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
143684 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction