Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM76B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178757
Description
FAM76B Polyclonal specifically detects FAM76B in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
FAM76B | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
family with sequence similarity 76, member B, hypothetical protein LOC143684 | |
Rabbit | |
Affinity Purified | |
RUO | |
143684 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 mcg/mg lysate | |
NP_653265.3 | |
FAM76B | |
The immunogen for this product maps to a region between residue 289 and 339 of human Family with Sequence Similarity 76, Member B using the numbering given in entry NP_653265.3 (GeneID 143684). | |
100 μL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction