Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM76B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557741 Supplier Invitrogen™ Supplier No. PA557741
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HHRHSSSHHK ISNLSPEEEQ GLWKQSHKSS ATIQNETPKK KPKLESKPSN GDSSSINQSA DSGGTDNFVL ISQLKEE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 61%.

FAM76B is an uncharacterized protein belonging to the FAM76 family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM76B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Fam76b
Gene Accession No. Q5HYJ3
Gene Alias 2810485I05Rik; C78303; FAM76B; family with sequence similarity 76 member B; family with sequence similarity 76, member B; protein FAM76B; RGD1311077
Gene Symbols Fam76b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM76B. Recombinant protein control fragment (Product #RP-97262).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143684
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.