Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM78A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554419
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554419 Supplier Invitrogen™ Supplier No. PA554419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FTTWLVATNT STNDMIILQT LHWRMQLSIE VNPNRPLGQR ARLREPIAQD QPKILSKNEP IP Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

FAM78A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM78A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM78A
Gene Accession No. Q5JUQ0
Gene Alias A130092J06Rik; C9orf59; chromosome 9 open reading frame 59; FAM78A; family with sequence similarity 78 member A; family with sequence similarity 78, member A; protein FAM78A; RGD1566351
Gene Symbols FAM78A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM78A. Recombinant protein control fragment (Product #RP-92946).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286336
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.