Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM78B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567281
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567281 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567281 Supplier Invitrogen™ Supplier No. PA567281
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IRRENIVVYDV CATIDQCPTR IEETSPIVLR YKTPYFKASA R Highest antigen sequence identity to the following orthologs - mouse 100%, rat 83%.

FAM78B (Family With Sequence Similarity 78 Member B) is a Protein Coding gene. An important paralog of this gene isFAM78A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM78B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM78B
Gene Accession No. Q5VT40
Gene Alias C030014K22Rik; C030020L09Rik; Fam78b; family with sequence similarity 78 member B; family with sequence similarity 78, member B; protein FAM78B
Gene Symbols FAM78B
Host Species Rabbit
Immunogen Recombinant Human FAM78B. Recombinant protein control fragment (Product #RP-107918).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149297
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.