Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM81A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566797
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566797 Supplier Invitrogen™ Supplier No. PA566797
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLSLIVKENSG ASERDMEKKL SQMSARLDKI EEGQKKTFDG QRTRQEEEKM HGRITKLELQ MNQNIKEMKA EVNAGFTAVY ESIGSLRQVL EAKMKLDRDQ LQKQIQ Highest antigen sequence identity to the following orthologs - mouse 81%, rat 83%.

FAM81A (Family With Sequence Similarity 81 Member A) is a Protein Coding gene. An important paralog of this gene isFAM81B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM81A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM81A
Gene Accession No. Q8TBF8
Gene Alias 6430514L14Rik; FAM81A; family with sequence similarity 81 member A; family with sequence similarity 81, member A; protein FAM81A; RGD1311958
Gene Symbols FAM81A
Host Species Rabbit
Immunogen Recombinant Human FAM81A. Recombinant protein control fragment (Product #RP-105370).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145773
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.