Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM81B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564476
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564476 Supplier Invitrogen™ Supplier No. PA564476
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MQLQFLGTLA SSEKRKKSQR LFFKNIKSTK NKAGKASIMS SDTNVNKSAS PTATAEEQPV EPDGPLPGSD NNQE Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 31%.

FAM81B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM81B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM81B
Gene Accession No. Q96LP2
Gene Alias FAM81B; family with sequence similarity 81 member B; family with sequence similarity 81, member B; Gm276; Protein FAM81B; RGD1561830
Gene Symbols FAM81B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM81B. Recombinant protein control fragment (Product #RP-104090).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153643
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.