Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM82A Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB404602 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB404602 0.025 mg
NBP241121 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB404602 Supplier Novus Biologicals Supplier No. NBP2411210.025MG
Only null left
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

FAM82A Polyclonal specifically detects FAM82A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FAM82A
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 5 -20 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias family with sequence similarity 82, member A1, member A, UNQ9371/PRO34163
Gene Symbols RMDN2
Host Species Chicken
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human FAM82A1. The immunogen is located within amino acids 140 - 190 of FAM82A1. Amino Acid Squence: ITANTDTEEQSFPVPK
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151393
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.