Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM82B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555321 Supplier Invitrogen™ Supplier No. PA555321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLARASRDVA QLSRTSEEEK KLLVYEALEY AKRALEKNES SFASHKWYAI CLSDVGDYEG IKAKIANAYI IKEHFEKAIE LNPKDATSIH L Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

FAM82B belongs to the FAM82/RMD family and contains two TPR repeats. The specific function of FAM82B is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM82B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RMDN1
Gene Accession No. Q96DB5
Gene Alias CGI-90; Fam82b; family with sequence similarity 82, member B; hRMD-1; microtubule-associated protein; Protein FAM82B; regulator of microtubule dynamics 1; regulator of microtubule dynamics protein 1; RMD1; RMD-1; RMDN1
Gene Symbols RMDN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM82B. Recombinant protein control fragment (Product #RP-93960).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51115
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.