Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM82B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598748
Description
FAM82B Polyclonal Antibody for Western Blot, IHC (P), ELISA
FAM82B belongs to the FAM82/RMD family and contains two TPR repeats. The specific function of FAM82B is not yet known.
Specifications
FAM82B | |
Polyclonal | |
Unconjugated | |
RMDN1 | |
CGI-90; Fam82b; family with sequence similarity 82, member B; hRMD-1; microtubule-associated protein; Protein FAM82B; regulator of microtubule dynamics 1; regulator of microtubule dynamics protein 1; RMD1; RMD-1; RMDN1 | |
Rabbit | |
Protein G | |
RUO | |
51115 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96DB5 | |
RMDN1 | |
Recombinant Human Regulator of microtubule dynamics protein 1 protein (13-314AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction