Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM82B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542837 Supplier Invitrogen™ Supplier No. PA542837
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MALAARLWRL LPFRRGAAPG SRLPAGTSGS RGHCGPCRFR GFEVMGNPGT Sequence homology: Human: 100%.

FAM82B belongs to the FAM82/RMD family and contains two TPR repeats. The specific function of FAM82B is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM82B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RMDN1
Gene Accession No. Q96DB5
Gene Alias CGI-90; Fam82b; family with sequence similarity 82, member B; hRMD-1; microtubule-associated protein; Protein FAM82B; regulator of microtubule dynamics 1; regulator of microtubule dynamics protein 1; RMD1; RMD-1; RMDN1
Gene Symbols RMDN1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FAM82B (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51115
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.