Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM83B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216423
Description
FAM83B Polyclonal specifically detects FAM83B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FAM83B | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
C6orf143, family with sequence similarity 83, member B, MGC126677, MGC138480 | |
Rabbit | |
115 kDa | |
0.1 mL | |
Primary | |
Expected to cross react based on sequence identity: Mouse (89%), Rat (89%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q5T0W9 | |
FAM83B | |
Recombinant protein encompassing a sequence within the N-terminus region of human FAM83B. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
222584 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction