Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM83B Mouse anti-Human, Unconjugated, Clone: 7D11, MilliporeSigma™

Catalog No. MABC112825 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1128-25 25 μg
MABC1128-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1128-25 Supplier MilliporeSigma Supplier No. MABC112825UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-FAM83B, clone 7D1, Cat. No. MABC1128, is a mouse monoclonal antibody that detects FAM83B and has been tested for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM83B
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7D11
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Protein FAM83B;Family with Sequence Similarity 83;member B
Gene Symbols FAM83B;Q5T0W9
Host Species Mouse
Immunogen His-tagged full length human recombinant Protein FAM83B.
Molecular Weight of Antigen ∽115 kDa observed; 114.80 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001010872
Test Specificity Clone 7D11 specifically detects human FAM83B protein.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.