Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM83C Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298186PCP
Description
FAM83C Polyclonal antibody specifically detects FAM83C in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
FAM83C | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
C20orf128, Chromosome 20 Open Reading Frame 128, dJ614O4.7, EC 1.6.5, EC 6.3.5.3, Family With Sequence Similarity 83, Member C, Protein FAM83C | |
Produced in rabbits immunized with E. coli-derived Human FAM83C fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
128876 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction