Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM83D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599011
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599011 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599011 Supplier Invitrogen™ Supplier No. PA599011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM83D Polyclonal Antibody for ELISA, IHC (P), Western Blot

Probable proto-oncogene that regulates cell proliferation, growth, migration and epithelial to mesenchymal transition. Through the degradation of FBXW7, may act indirectly on the expression and downstream signaling of MTOR, JUN and MYC (PubMed:24344117). May play also a role in cell proliferation through activation of the ERK1/ERK2 signaling cascade (PubMed:25646692). May also be important for proper chromosome congression and alignment during mitosis through its interaction with KIF22.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM83D
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FAM83D
Gene Accession No. Q9H4H8
Gene Alias 2310007D09Rik; BB104611; C20orf129; CHICA; dJ616B8.3; FAM83D; family with sequence similarity 83 member D; family with sequence similarity 83, member D; mitotic spindle protein CHICA; protein FAM83D; RGD1565583; spindle protein CHICA
Gene Symbols FAM83D
Host Species Rabbit
Immunogen Recombinant Human Protein FAM83D protein (339-470AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81610
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.