Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM83D Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140100
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ASSTGSPASI RTTDFHNPGY PKYLGTPHLE LYLSDSLRNL NKERQFHFAG IRSRLNHMLA MLSRRTLFTE NHLGLHSGNF S.
Probable proto-oncogene that regulates cell proliferation, growth, migration and epithelial to mesenchymal transition. Through the degradation of FBXW7, may act indirectly on the expression and downstream signaling of MTOR, JUN and MYC (PubMed:24344117). May play also a role in cell proliferation through activation of the ERK1/ERK2 signaling cascade (PubMed:25646692). May also be important for proper chromosome congression and alignment during mitosis through its interaction with KIF22.
Specifications
FAM83D | |
Polyclonal | |
Unconjugated | |
FAM83D | |
2310007D09Rik; BB104611; C20orf129; CHICA; dJ616B8.3; FAM83D; family with sequence similarity 83 member D; family with sequence similarity 83, member D; mitotic spindle protein CHICA; protein FAM83D; RGD1565583; spindle protein CHICA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81610 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H4H8 | |
FAM83D | |
Recombinant protein corresponding to Human FAM83D. Recombinant protein control fragment (Product #RP-108985). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction