Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM83D Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571098
Description
This target displays homology in the following species: Horse: 79%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 79%.
Probable proto-oncogene that regulates cell proliferation, growth, migration and epithelial to mesenchymal transition. Through the degradation of FBXW7, may act indirectly on the expression and downstream signaling of MTOR, JUN and MYC (PubMed:24344117). May play also a role in cell proliferation through activation of the ERK1/ERK2 signaling cascade (PubMed:25646692). May also be important for proper chromosome congression and alignment during mitosis through its interaction with KIF22.
Specifications
FAM83D | |
Polyclonal | |
Unconjugated | |
FAM83D | |
2310007D09Rik; BB104611; C20orf129; CHICA; dJ616B8.3; FAM83D; family with sequence similarity 83 member D; family with sequence similarity 83, member D; mitotic spindle protein CHICA; protein FAM83D; RGD1565583; spindle protein CHICA | |
Rabbit | |
Affinity chromatography | |
RUO | |
81610 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H4H8 | |
FAM83D | |
synthetic peptide directed towards the C-terminal region of human FAM83D. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction