Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM83E Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570272
Description
This target displays homology in the following species: Cow: 93%; Dog: 92%; Guinea Pig: 86%; Human: 100%; Mouse: 86%; Pig: 100%; Rat: 86%.
FAM83E may play a role in MAPK signaling.
Specifications
FAM83E | |
Polyclonal | |
Unconjugated | |
FAM83E | |
4930403C10Rik; FAM83E; family with sequence similarity 83 member E; family with sequence similarity 83, member E; protein FAM83E; RGD1309316 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54854 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q2M2I3 | |
FAM83E | |
synthetic peptide directed towards the middle region of human FAM83E. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction