Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM83E Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145122 Supplier Invitrogen™ Supplier No. PA5145122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DNELKKSWGS KDTPAKALMR QRGTGGGPWG EVDSRPPWGG ALPLPPAHRL RYLSPARRRF GGDATFKLQE P Antigen sequence identity: Mouse 67%; Rat 65%.

FAM83E may play a role in MAPK signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM83E
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM83E
Gene Accession No. Q2M2I3
Gene Alias 4930403C10Rik; FAM83E; family with sequence similarity 83 member E; family with sequence similarity 83, member E; protein FAM83E; RGD1309316
Gene Symbols FAM83E
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 390-460. Recombinant protein control fragment (Product #RP-109872).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54854
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.