Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM83E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561928
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561928 Supplier Invitrogen™ Supplier No. PA561928
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPVYLLLDRQ QLPAFLELAQ QLGVNPWNTE NVDVRVVRGC SFQSRWRRQV SGTVREKFVL LDGERVISGS YS Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.

FAM83E may play a role in MAPK signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM83E
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM83E
Gene Accession No. Q2M2I3
Gene Alias 4930403C10Rik; FAM83E; family with sequence similarity 83 member E; family with sequence similarity 83, member E; protein FAM83E; RGD1309316
Gene Symbols FAM83E
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM83E. Recombinant protein control fragment (Product #RP-98984).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54854
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.