Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM83E Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983941
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3941 Supplier Bioss Supplier No. BS16016R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM83E may play a role in MAPK signaling.

Specifications

Antigen FAM83E
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FAM83E
Gene Accession No. Q2M2I3
Gene Alias 4930403C10Rik; FAM83E; family with sequence similarity 83 member E; family with sequence similarity 83, member E; Protein FAM83E; RGD1309316
Gene Symbols FAM83E
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM83E.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54854
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.