Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    FAM83F Antibody, DyLight 680, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305913FR
Description
FAM83F Polyclonal specifically detects FAM83F in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FAM83F | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| family with sequence similarity 83, member F, hypothetical protein LOC113828 | |
| Produced in rabbits immunized with E. coli-derived Human FAM83F fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified | 
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 113828 | |
| Store at 4°C in the dark. | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            