Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM83F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566703
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566703 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566703 Supplier Invitrogen™ Supplier No. PA566703
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SYRFTWSSSHV DRNLLLLLTG QNVEPFDTEF RELYAISEEV DLYRQLSLAG RVGLHYSSTV ARKLIN Highest antigen sequence identity to the following orthologs - mouse 82%, rat 85%.

FAM83F is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM83F
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM83F
Gene Accession No. Q8NEG4
Gene Alias AW544981; FAM83F; family with sequence similarity 83 member F; family with sequence similarity 83, member F; protein FAM83F; RGD1562089
Gene Symbols FAM83F
Host Species Rabbit
Immunogen Recombinant Human FAM83F. Recombinant protein control fragment (Product #RP-107354).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113828
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.