Learn More
FAM83G Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304282AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
May regulate the bone morphogenetic proteins (BMP) pathway.Specifications
FAM83G | |
Polyclonal | |
Unconjugated | |
FAM83G | |
Protein FAM83G, Protein associated with SMAD1, FAM83G, PAWS1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
644815 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
A6ND36 | |
FAM83G | |
Between 773 and 823 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.