Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM84A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561571
Description
Immunogen sequence: PSGIEKDELR VGVAYFFSDD EEDLDERGQP DKFGVKAPPG CTPCPESPSR HHHHLLHQLV LNETQFSAFR GQECIFSKVS GGP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Specifications
FAM84A | |
Polyclonal | |
Unconjugated | |
FAM84A | |
2310003N02Rik; 4731402F03Rik; AW125753; Fam84a; family with sequence similarity 84 member A; family with sequence similarity 84, member A; LRAT domain-containing 1; LRATD1; Neurologic sensory protein 1; neurological/sensory 1; Nse1; PP11517; protein FAM84A; Protein LRATD1; RGD1305779 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
151354 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96KN4 | |
FAM84A | |
Recombinant protein corresponding to Human FAM84A. Recombinant protein control fragment (Product #RP-98523). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction