Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM84A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530705
Description
Recommended positive controls: IMR32. Predicted reactivity: Human (99%), Mouse (95%), Rat (95%), Rhesus Monkey (99%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Specifications
FAM84A | |
Polyclonal | |
Unconjugated | |
FAM84A | |
2310003N02Rik; 4731402F03Rik; AW125753; Fam84a; family with sequence similarity 84 member A; family with sequence similarity 84, member A; LRAT domain-containing 1; LRATD1; Neurologic sensory protein 1; neurological/sensory 1; Nse1; PP11517; protein FAM84A; Protein LRATD1; RGD1305779 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
151354 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96KN4 | |
FAM84A | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 242 of Human FAM84A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction