Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM84B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566458
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566458 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566458 Supplier Invitrogen™ Supplier No. PA566458
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGNQVEKLTHL SYKEVPTADP TGVDRDDGPR IGVSYIFSND DEDVEPQPPP QGPDGGGLPD GGDGPPPPQP QPYD Highest antigen sequence identity to the following orthologs - mouse 91%, rat 89%.

FAM84B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM84B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM84B
Gene Accession No. Q96KN1
Gene Alias BCMP101; BF642811; breast cancer membrane protein 101; breast cancer membrane-associated protein 101; D330050I23Rik; FAM84B; family with sequence similarity 84 member B; family with sequence similarity 84, member B; LRAT domain-containing 2; LRATD2; neurological/sensory 2; NSE2; Protein FAM84B; Protein LRATD2; Protein NSE2; RGD1562236
Gene Symbols FAM84B
Host Species Rabbit
Immunogen Recombinant Human FAM84B. Recombinant protein control fragment (Product #RP-107132).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157638
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.