Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM86A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113282
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113282 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113282 Supplier Invitrogen™ Supplier No. PA5113282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM86A Polyclonal Antibody for ELISA, IHC (P), Western Blot

Catalyzes the trimethylation of eukaryotic elongation factor 2 (EEF2) on 'Lys-525'. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM86A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EEF2KMT
Gene Accession No. Q96G04
Gene Alias EEF2KMT; eEF2-KMT; eEF2-lysine methyltransferase; eukaryotic elongation factor 2 lysine methyltransferase; FAM86A; family with sequence similarity 86, member A; protein FAM86A; protein-lysine N-methyltransferase EEF2KMT; putative protein N-methyltransferase FAM86A; SB153
Gene Symbols EEF2KMT
Host Species Rabbit
Immunogen Recombinant Human Protein-lysine N-methyltransferase EEF2KMT protein (1-330AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196483
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.