Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM90A1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299202MFV610
Description
FAM90A1 Polyclonal antibody specifically detects FAM90A1 in Human samples. It is validated for Western BlotSpecifications
FAM90A1 | |
Polyclonal | |
Western Blot | |
family with sequence similarity 90, member A1, FLJ10408, hypothetical protein LOC55138 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human FAM90A1. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
55138 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction