Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM90A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139847
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139847 Supplier Invitrogen™ Supplier No. PA5139847
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: APIQACLNFP KKPRLGPFQI PESAIQGGEL GAPENLQPPP AATELGP Highest antigen sequence indentity to the following orthologs: Mouse - 36%, Rat - 36%.

FAM90A1 belongs to subfamily I of the primate-specific FAM90A gene family, which originated from multiple duplications and rearrangements (Bosch et al., 2007 [PubMed 17684299]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM90A1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM90A1
Gene Accession No. Q86YD7
Gene Alias FAM90A1; family with sequence similarity 90 member A1; protein FAM90A1
Gene Symbols FAM90A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM90A1. Recombinant protein control fragment (Product #RP-108978).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55138
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.