Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM90A12P Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571099
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571099 Supplier Invitrogen™ Supplier No. PA571099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

FAM90A12 belongs to subfamily II of the primate-specific FAM90A gene family, which originated from multiple duplications and rearrangements. For background information on the FAM90A gene family, as well as information on the evolution of FAM90A genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM90A12P
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM90A12P
Gene Accession No. A8MX19
Gene Symbols FAM90A12P
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FAM90A12P.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 645879
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.