Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM91A1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079831
Description
FAM91A1 Polyclonal specifically detects FAM91A1 in Human samples. It is validated for Immunoprecipitation.Specifications
FAM91A1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
DKFZp666B104, family with sequence similarity 91, member A1, FLJ23790, hypothetical protein LOC157769, skeletal muscle cells re-entry induced | |
Rabbit | |
Affinity Purified | |
RUO | |
157769 | |
Store at 4C. Do not freeze. |
Immunoprecipitation | |
1.0 mg/mL | |
Immunoprecipitation 2 - 5 ug/mg lysate | |
NP_659400.2 | |
FAM91A1 | |
The immunogen recognized by this antibody maps to a region between residue 650 and 700 of human family with sequence similarity 91, member A1 using the numbering given in entry NP_659400.2 (GeneID 157769). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction