Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM91A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546446
Description
Peptide sequence: RHNYPWNKLP ANVRQSLGNS QREYEKQVVL YSIRNQLRYR NNLVKHVKKD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
FAM91A1 (family with sequence similarity 91, member A1) is a 94 kDa phospho-protein that is not yet characterized.
Specifications
FAM91A1 | |
Polyclonal | |
Unconjugated | |
FAM91A1 | |
AV220772; BC033609; D15Ertd621e; DNA segment, Chr 15, ERATO Doi 621, expressed; Fam91a1; family with sequence similarity 91 member A1; family with sequence similarity 91, member A1; hypothetical protein FLJ23790; hypothetical protein LOC393413; Kiaa0493; mKIAA0493; Protein FAM91A1; si:dkey-5e12.2; skeletal muscle cells re-entry induced; SKIN; zgc:63609 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
157769 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q658Y4 | |
FAM91A1 | |
Synthetic peptide directed towards the N-terminal of human FAM91A1 (aa 10-59). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction