Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM91A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555741 Supplier Invitrogen™ Supplier No. PA555741
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHGIGETVHV PFPFDETELQ GEFTRVNMGV HKALQILRNR VDLQHLCGYV TMLNASSQLA DRKLSDASDE RGEPDLASGS Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

FAM91A1 (family with sequence similarity 91, member A1) is a 94 kDa phospho-protein that is not yet characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM91A1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM91A1
Gene Accession No. Q658Y4
Gene Alias AV220772; BC033609; D15Ertd621e; DNA segment, Chr 15, ERATO Doi 621, expressed; Fam91a1; family with sequence similarity 91 member A1; family with sequence similarity 91, member A1; hypothetical protein FLJ23790; hypothetical protein LOC393413; Kiaa0493; mKIAA0493; Protein FAM91A1; si:dkey-5e12.2; skeletal muscle cells re-entry induced; SKIN; zgc:63609
Gene Symbols FAM91A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM91A1. Recombinant protein control fragment (Product #RP-93961).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157769
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.