Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM96A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566628
Description
Immunogen sequence: LATLIGLCLRV KLQRCLPFKH KLEIYISEGT HSTEEDINKQ INDKERVAAA Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Component of the cytosolic iron-sulfur protein assembly (CIA) complex, a multiprotein complex that mediates the incorporation of iron-sulfur cluster into extramitochondrial Fe/S proteins (PubMed:23891004). As a CIA complex component and in collaboration with CIAO1 specifically matures ACO1 and stabilizes IREB2, connecting cytosolic iron-sulfur protein maturation with cellular iron regulation (PubMed:23891004). May play a role in chromosome segregation through establishment of sister chromatid cohesion. May induce apoptosis in collaboration with APAF1 (PubMed:25716227). [UniProt].
Specifications
FAM96A | |
Polyclonal | |
Unconjugated | |
FAM96A | |
5730536A07Rik; CIA2A; Ciao2a; Cytosolic iron-sulfur assembly component 2A; Fam96a; family with sequence similarity 96 member A; family with sequence similarity 96, member A; MIP18 family protein FAM96A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84191 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H5X1 | |
FAM96A | |
Recombinant Human FAM96A. Recombinant protein control fragment (Product #RP-105277). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction