Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM96A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559187
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559187 Supplier Invitrogen™ Supplier No. PA559187
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RIMEEKALEV YDLIRTIRDP EKPNTLEELE VVSESCVEVQ EINEEEYLVI IRFTPTVPHC SL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.

Component of the cytosolic iron-sulfur protein assembly (CIA) complex, a multiprotein complex that mediates the incorporation of iron-sulfur cluster into extramitochondrial Fe/S proteins (PubMed:23891004). As a CIA complex component and in collaboration with CIAO1 specifically matures ACO1 and stabilizes IREB2, connecting cytosolic iron-sulfur protein maturation with cellular iron regulation (PubMed:23891004). May play a role in chromosome segregation through establishment of sister chromatid cohesion. May induce apoptosis in collaboration with APAF1 (PubMed:25716227). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM96A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM96A
Gene Accession No. Q9H5X1
Gene Alias 5730536A07Rik; CIA2A; Ciao2a; Cytosolic iron-sulfur assembly component 2A; Fam96a; family with sequence similarity 96 member A; family with sequence similarity 96, member A; MIP18 family protein FAM96A
Gene Symbols FAM96A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM96A. Recombinant protein control fragment (Product #RP-98168).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84191
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.