Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM96B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596815
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596815 Supplier Invitrogen™ Supplier No. PA596815
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVGGGGVGGG LLENANPLIY QRSGERPVTA GEEDEQVPDS IDAREIFDLI RSINDPEHPL TLEELNVVEQ VRVQVSDPES TVAVAFTPTI PHCSMATLIG LSIKVKLLRS LPQRFKMDVH ITPGTHASEH AVNKQLADKE RVAAALENTH LLEVVNQCLS ARS; Positive Samples: HeLa, NCI-H460, MCF-7, Mouse heart, Mouse testis; Cellular Location: Cytoplasm, Nucleus, cytoskeleton, spindle.

Signs of Rubinstein-Taybi include mental retardation and predisposition to tumor growth and white blood cell neoplasias. Crohn's disease is a gastrointestinal inflammatory condition associated with chromosome 16 through the NOD2 gene. An association with systemic lupus erythematosis and a number of other autoimmune disorders with the pericentromeric region of chromosome 16 has led to the identification of SLC5A11 as a potential autoimmune modifier. The FAM96B gene product has been provisionally designated FAM96B pending further characterization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM96B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CIAO2B
Gene Accession No. Q9D187, Q9Y3D0
Gene Alias 1110019N10Rik; CGI-128; CIAB; Ciao2b; Cytosolic iron-sulfur assembly component 2B; Fam96b; family with sequence similarity 96 member B; family with sequence similarity 96, member B; HSPC118; MIP18; Mitotic spindle-associated MMXD complex subunit MIP18; MSS19-interacting protein of 18 kDa; Protein FAM96B; RGD1311762
Gene Symbols CIAO2B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-163 of human FAM96B (NP_0571461).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51647, 68523
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.